Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUDT14
Entrez Gene ID
397008
Gene Name
nudix hydrolase 14
Gene Aliases
UGPP
Chromosome Location
Chr.Unknown: 3289576 - 3296920 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_021081551.1 XP_020937210.1 - 540 111
GenBank mRNA AJ539380.1 - - 435 111

View Full Product Details