Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZAR1
Entrez Gene ID
574050
Gene Name
zygote arrest 1
Gene Aliases
-
Chromosome Location
Chr.8: 38386520 - 38391618 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129956.1 NP_001123428.1 2 - 3 938 64
GenBank mRNA AM040194.1 - 1 - 2 59 64
DQ231444.1 - 2 - 3 938 64
DQ231445.1 - 2 - 3 392 64
DQ231446.1 - 3 - 4 245 64
DQ231447.1 - 3 - 4 578 64

View Full Product Details