Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPAM
Entrez Gene ID
397629
Gene Name
glycerol-3-phosphate acyltransferase, mitochondrial
Gene Aliases
GPAT, GPAT-1, GPAT1
Chromosome Location
Chr.14: 122538600 - 122582390 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005671462.3 XP_005671519.2 - 5311 108
GenBank mRNA AK348444.1 - 17 - 18 2218 108
AY284842.1 - 3 - 4 385 108

View Full Product Details