Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RXRG
Entrez Gene ID
445465
Gene Name
retinoid X receptor gamma
Gene Aliases
NR2B3, RXRgamma
Chromosome Location
Chr.4: 85158313 - 85357116 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130213.1 NP_001123685.1 7 - 8 1217 74
XM_005655370.3 XP_005655427.1 - 993 74
XM_013988076.2 XP_013843530.1 - 1875 74
XM_021088737.1 XP_020944396.1 - 1270 74
GenBank mRNA AB530150.1 - 5 - 6 678 74
AK237831.1 - 6 - 7 973 74
AK394594.1 - 6 - 7 978 74
AY429474.1 - 6 - 7 952 74
DQ866834.1 - 7 - 8 1217 74

View Full Product Details