Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DMP1
Entrez Gene ID
399688
Gene Name
dentin matrix acidic phosphoprotein 1
Gene Aliases
-
Chromosome Location
Chr.8: 131285238 - 131319398 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129953.2 NP_001123425.1 4 - 5 263 83
XM_005667009.3 XP_005667066.1 - 2024 83
XM_005667010.3 XP_005667067.1 - 2009 83
XM_021100481.1 XP_020956140.1 - 1274 83
XM_021100482.1 XP_020956141.1 - 911 83
XM_021100483.1 XP_020956142.1 - 3338 83
GenBank mRNA AY524986.1 - 3 - 4 108 83
AY963261.1 - 4 - 5 263 83

View Full Product Details