Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS4X
Entrez Gene ID
397611
Gene Name
ribosomal protein S4 X-linked
Gene Aliases
RPS4
Chromosome Location
Chr.X: 58149302 - 58155179 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204283.1 NP_001191212.1 4 - 5 404 117
GenBank mRNA AK232196.1 - 4 - 5 404 117
AK233807.1 - 4 - 5 378 117
AK394418.1 - 4 - 5 411 117
D50110.1 - 3 - 4 298 117

View Full Product Details