Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- POU3F1
- Entrez Gene ID
-
397513
- Gene Name
- POU class 3 homeobox 1
- Gene Aliases
-
BRN1, POU3F3
- Chromosome Location
-
Chr.6: 94025133 - 94028053 on Build Sscrofa11.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
L28764.1 |
- |
1 - 1 |
246 |
80 |
- Gene Symbol
- POU3F3
- Entrez Gene ID
-
100737124
- Gene Name
- POU class 3 homeobox 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 49804112 - 49807730 on Build Sscrofa11.1
View Full Product Details