Assay Design
The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Gene Transcripts
- Gene Symbol
- IGH
- Entrez Gene ID
-
406179
- Gene Name
- immunoglobulin heavy locus
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U15194.1 |
- |
- |
339 |
76 |
View Full Product Details