Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
GPD2
Entrez Gene ID
397348
Gene Name
glycerol-3-phosphate dehydrogenase 2
Gene Aliases
G3PD, MGPD
Chromosome Location
Chr.15: 63592723 - 63728636 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_021075775.1 XP_020931434.1 - 2333 89
XM_021075776.1 XP_020931435.1 - 2333 89
XM_021075777.1 XP_020931436.1 - 2093 89
XM_021075778.1 XP_020931437.1 - 2093 89
XM_021075779.1 XP_020931438.1 - 2161 89
XM_021075780.1 XP_020931439.1 - 2115 89
GenBank mRNA AF141957.1 - 4 - 4 352 89
AK344046.1 - 16 - 16 2012 89

View Full Product Details