Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
SLC9A5
Entrez Gene ID
396753
Gene Name
solute carrier family 9 member A5
Gene Aliases
NHE5
Chromosome Location
Chr.6: 27883163 - 27904076 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_021094077.1 XP_020949736.1 - 2002 102
XM_021094079.1 XP_020949738.1 - 2000 102
XM_021094080.1 XP_020949739.1 - 2001 102
GenBank mRNA AF260664.1 - 2 - 2 152 102
AK392094.1 - 3 - 3 575 102

View Full Product Details