Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
INSIG1
Entrez Gene ID
403162
Gene Name
insulin induced gene 1
Gene Aliases
CL-6
Chromosome Location
Chr.18: 2885494 - 2898758 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY336601.2 - 1 - 1 56 133

View Full Product Details