Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CYP3A46
Entrez Gene ID
100126281
Gene Name
cytochrome P450 3A46
Gene Aliases
CYP3A39, CYP3A46v3, CYP3A88
Chromosome Location
Chr.3: 6677911 - 6709855 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134824.1 NP_001128296.1 13 - 13 1673 92
GenBank mRNA AK389809.1 - 13 - 13 1701 92
EF625347.2 - 13 - 13 1673 92

View Full Product Details