Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS28
Entrez Gene ID
414420
Gene Name
ribosomal protein S28
Gene Aliases
-
Chromosome Location
Chr.2: 70953706 - 70954862 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001587.1 NP_001001587.1 3 - 3 156 66
XM_013987364.2 XP_013842818.1 - 241 66
GenBank mRNA AK235060.1 - 3 - 3 154 66
AK391755.1 - 3 - 3 155 66
AY550038.1 - 3 - 3 156 66

View Full Product Details