Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
IL27
Entrez Gene ID
493187
Gene Name
interleukin 27
Gene Aliases
IL-27p28, IL27A
Chromosome Location
Chr.3: 18421962 - 18427916 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007520.1 NP_001007521.1 5 - 5 549 122
GenBank mRNA AK345589.1 - 5 - 5 597 122
AY788913.1 - 5 - 5 549 122

View Full Product Details