Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AQN-1
Entrez Gene ID
396829
Gene Name
AQN-1 protein
Gene Aliases
-
Chromosome Location
Chr.14: 132258948 - 132266059 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025210.1 NP_001020381.1 4 - 4 357 85
XM_005655814.2 XP_005655871.1 - 471 85
GenBank mRNA AJ004998.1 - 2 - 2 263 85
AJ853854.1 - 4 - 4 357 85
AJ853855.1 - 4 - 4 357 85

View Full Product Details