Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
GALP
Entrez Gene ID
396772
Gene Name
galanin like peptide
Gene Aliases
-
Chromosome Location
Chr.6: 60485612 - 60493591 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001348949.1 NP_001335878.1 3 - 3 231 110
NM_213825.2 NP_998990.1 3 - 3 231 110
GenBank mRNA AF188490.1 - 2 - 2 144 110
AK348630.1 - 3 - 3 194 110
AW436648.1 - 3 - 3 232 110
BP455542.1 - 3 - 3 190 110
FD607412.1 - 3 - 3 196 110

View Full Product Details