Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CBR2
Entrez Gene ID
396780
Gene Name
carbonyl reductase 2
Gene Aliases
-
Chromosome Location
Chr.12: 973780 - 976017 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213827.1 NP_998992.1 8 - 8 651 64
GenBank mRNA AK231874.1 - 8 - 8 671 64
AK345555.1 - 8 - 8 668 64
AK345737.1 - 8 - 8 733 64
D16511.1 - 8 - 8 651 64

View Full Product Details