Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
FSHB
Entrez Gene ID
396895
Gene Name
follicle stimulating hormone subunit beta
Gene Aliases
FSH-beta
Chromosome Location
Chr.2: 30395769 - 30399282 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213875.1 NP_999040.1 3 - 3 251 70
GenBank mRNA AF134151.1 - 3 - 3 251 70
AK349340.1 - 3 - 3 254 70
M35676.1 - 2 - 2 138 70

View Full Product Details