Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEOX2
Entrez Gene ID
397222
Gene Name
mesenchyme homeobox 2
Gene Aliases
GAX
Chromosome Location
Chr.9: 84945178 - 85009939 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214121.1 NP_999286.1 3 - 3 851 65
GenBank mRNA AF295686.1 - 3 - 3 851 65

View Full Product Details