Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BIRC3
- Entrez Gene ID
-
100622590
- Gene Name
- baculoviral IAP repeat containing 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 33003570 - 33020252 on Build Sscrofa11.1
- Gene Symbol
- PIAP
- Entrez Gene ID
-
397374
- Gene Name
- inhibitor of apoptosis-like
- Gene Aliases
-
BIRC2
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U79142.1 |
- |
7 - 7 |
1055 |
89 |
View Full Product Details