Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
FXYD3
Entrez Gene ID
397413
Gene Name
FXYD domain containing ion transport regulator 3
Gene Aliases
MAT-8
Chromosome Location
Chr.6: 44647771 - 44654377 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214208.2 NP_999373.1 4 - 4 248 114
XM_005664469.3 XP_005664526.2 - 399 114
GenBank mRNA AB015759.1 - 4 - 4 249 114
AK350001.1 - 5 - 5 298 114

View Full Product Details