Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
AREG
Entrez Gene ID
397668
Gene Name
amphiregulin
Gene Aliases
-
Chromosome Location
Chr.8: 70417935 - 70428106 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214376.1 NP_999541.1 5 - 5 847 92
GenBank mRNA AY028310.1 - 5 - 5 847 92

View Full Product Details