Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELSPBP1
Entrez Gene ID
399529
Gene Name
epididymal sperm binding protein 1
Gene Aliases
E12
Chromosome Location
Chr.6: 53583965 - 53603746 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214404.1 NP_999569.1 5 - 5 550 71
XM_021093579.1 XP_020949238.1 - 448 71
GenBank mRNA AJ539176.1 - 5 - 5 550 71

View Full Product Details