Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
NDUFA4
Entrez Gene ID
100579141
Gene Name
NDUFA4 mitochondrial complex associated
Gene Aliases
-
Chromosome Location
Chr.9: 80637005 - 80644067 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097468.2 NP_001090937.1 2 - 2 153 87
GenBank mRNA DQ629144.1 - 2 - 2 154 87
DY843591.1 - 2 - 2 154 87

View Full Product Details