Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
S1PR5
Entrez Gene ID
100141401
Gene Name
sphingosine-1-phosphate receptor 5
Gene Aliases
EDG8, S1P5
Chromosome Location
Chr.2: 69293537 - 69299408 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123109.1 NP_001116581.1 1 - 1 1047 136
XM_021076722.1 XP_020932381.1 - 1649 136
XM_021076730.1 XP_020932389.1 - 1143 136
XM_021076733.1 XP_020932392.1 - 1089 136
XM_021076735.1 XP_020932394.1 - 2432 136
XM_021076745.1 XP_020932404.1 - 1195 136
XM_021076747.1 XP_020932406.1 - 1494 136
XM_021076755.1 XP_020932414.1 - 1372 136
GenBank mRNA AK395472.1 - 2 - 2 1174 136

View Full Product Details