Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS3
Entrez Gene ID
733671
Gene Name
ribosomal protein S3
Gene Aliases
-
Chromosome Location
Chr.9: 9612939 - 9630404 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044601.1 NP_001038066.1 3 - 4 256 138
XM_021102267.1 XP_020957926.1 - 774 138
XM_021102268.1 XP_020957927.1 - 682 138
XM_021102269.1 XP_020957928.1 - 682 138
GenBank mRNA AK234402.1 - 5 - 6 755 138
AK390990.1 - 5 - 6 688 138
AK392066.1 - 3 - 4 282 138
AK398968.1 - 3 - 4 299 138
DQ660373.1 - 3 - 4 256 138

View Full Product Details