Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTBP3
Entrez Gene ID
768100
Gene Name
polypyrimidine tract binding protein 3
Gene Aliases
ROD1
Chromosome Location
Chr.1: 252986716 - 253093059 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077215.1 NP_001070683.1 11 - 12 1361 66
XM_005660329.3 XP_005660386.1 - 1629 66
XM_013993751.2 XP_013849205.1 - 1665 66
XM_013993752.2 XP_013849206.1 - 1600 66
XM_013993753.2 XP_013849207.1 - 1593 66
XM_021084313.1 XP_020939972.1 - 1584 66
GenBank mRNA AK236448.1 - 12 - 13 1569 66
AK343731.1 - 11 - 12 1522 66
DQ972969.1 - 11 - 12 1361 66

View Full Product Details