Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DTNBP1
Entrez Gene ID
100049697
Gene Name
dystrobrevin binding protein 1
Gene Aliases
-
Chromosome Location
Chr.7: 11602470 - 11720156 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098600.1 NP_001092070.1 2 - 3 156 87
GenBank mRNA AB271940.1 - 2 - 3 156 87
AK346417.1 - 2 - 3 233 87

View Full Product Details