Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BPIFB5
Entrez Gene ID
100113428
Gene Name
long palate lung and nasal epithelium protein 5
Gene Aliases
-
Chromosome Location
Chr.17: 36811645 - 36835958 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101033.1 NP_001094503.1 7 - 8 774 70
XM_005672829.3 XP_005672886.1 - 2273 70
XM_021077174.1 XP_020932833.1 - 2273 70
GenBank mRNA AK237864.1 - 7 - 8 780 70
AK350647.1 - 7 - 8 791 70
DQ859019.1 - 7 - 8 774 70

View Full Product Details