Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPIFB5
Entrez Gene ID
100113428
Gene Name
long palate lung and nasal epithelium protein 5
Gene Aliases
-
Chromosome Location
Chr.17: 36811645 - 36835958 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101033.1 NP_001094503.1 9 - 10 1049 79
XM_005672829.3 XP_005672886.1 - 2548 79
XM_021077174.1 XP_020932833.1 - 2548 79
GenBank mRNA AK237864.1 - 9 - 10 1055 79
AK350647.1 - 9 - 10 1066 79
DQ859019.1 - 9 - 10 1049 79

View Full Product Details