Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD1D
Entrez Gene ID
100124526
Gene Name
CD1d molecule
Gene Aliases
CD1
Chromosome Location
Chr.4: 92096670 - 92101177 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102680.2 NP_001096150.2 4 - 5 959 69
XM_021088435.1 XP_020944094.1 - 936 69
XM_021088436.1 XP_020944095.1 - 885 69
GenBank mRNA AB189911.1 - 4 - 5 917 69
AK238636.1 - 4 - 5 951 69
AK238943.1 - 3 - 4 1258 69
AK351633.1 - 2 - 3 389 69
AK352027.1 - 4 - 5 943 69
AK394569.1 - 4 - 5 937 69

View Full Product Details