Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RETNLB
Entrez Gene ID
100037943
Gene Name
resistin like beta
Gene Aliases
-
Chromosome Location
Chr.13: 150716863 - 150718405 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103210.1 NP_001096680.1 1 - 2 125 123
XM_005670236.3 XP_005670293.1 - 244 123
GenBank mRNA EU086593.1 - 1 - 2 125 123

View Full Product Details