Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNCG
Entrez Gene ID
100125343
Gene Name
synuclein gamma
Gene Aliases
-
Chromosome Location
Chr.14: 87916353 - 87922111 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103211.1 NP_001096681.1 3 - 4 340 106
XM_013983396.2 XP_013838850.1 - 764 106
XM_013983397.2 XP_013838851.1 - 472 106
GenBank mRNA AK236982.1 - 3 - 4 397 106
AK348791.1 - 3 - 4 411 106
AK348968.1 - 3 - 4 413 106
AK391883.1 - 3 - 4 414 106
EF104640.1 - 3 - 4 340 106

View Full Product Details