Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IFNE
Entrez Gene ID
100125969
Gene Name
interferon epsilon
Gene Aliases
IFN-epsilon, IFNE1
Chromosome Location
Chr.1: 201219848 - 201228361 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105310.1 NP_001098780.1 1 - 1 451 85
XM_005660060.3 XP_005660117.1 - 4202 85
GenBank mRNA EU072421.1 - 1 - 1 451 85
GQ415085.1 - 1 - 1 451 85

View Full Product Details