Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMO4
Entrez Gene ID
100127155
Gene Name
LIM domain only 4
Gene Aliases
-
Chromosome Location
Chr.4: 128894113 - 128910648 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112686.1 NP_001106156.1 3 - 4 464 68
XM_021088448.1 XP_020944107.1 - 792 68
XM_021088449.1 XP_020944108.1 - 482 68
GenBank mRNA AB304401.1 - 3 - 4 464 68
AK233224.1 - 3 - 4 702 68
AK235142.1 - 2 - 3 600 68
AK390167.1 - 3 - 4 704 68
AK390242.1 - 2 - 3 585 68
AK390347.1 - 3 - 4 699 68
AK390590.1 - 2 - 3 609 68
AY610416.1 - 3 - 4 392 68

View Full Product Details