Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPL5
Entrez Gene ID
100135671
Gene Name
ribosomal protein L5
Gene Aliases
-
Chromosome Location
Chr.4: 124359786 - 124367612 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114284.2 NP_001107756.1 3 - 4 231 147
GenBank mRNA AK237194.1 - - 597 147
AK350182.1 - 3 - 4 289 147
AK391704.1 - 3 - 4 264 147
AK393844.1 - 3 - 4 263 147
AK394513.1 - 3 - 4 269 147
EU352798.1 - 3 - 4 232 147

View Full Product Details