Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL5
Entrez Gene ID
100135671
Gene Name
ribosomal protein L5
Gene Aliases
-
Chromosome Location
Chr.4: 124359786 - 124367612 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114284.2 NP_001107756.1 6 - 7 756 141
GenBank mRNA AK350182.1 - 6 - 7 814 141
AK391704.1 - 6 - 7 789 141
AK393844.1 - 6 - 7 788 141
AK398700.1 - 6 - 7 790 141
EU352798.1 - 6 - 7 757 141

View Full Product Details