The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114284.2 | NP_001107756.1 | 7 - 8 | 837 | 94 |
GenBank mRNA | AK237194.1 | - | - | 1200 | 94 |
AK350182.1 | - | 7 - 8 | 895 | 94 | |
AK391704.1 | - | 7 - 8 | 870 | 94 | |
AK393844.1 | - | 7 - 8 | 869 | 94 | |
AK398700.1 | - | 7 - 8 | 871 | 94 | |
EU352798.1 | - | 7 - 8 | 838 | 94 |