Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPCS1
Entrez Gene ID
100135675
Gene Name
signal peptidase complex subunit 1
Gene Aliases
SPC12
Chromosome Location
Chr.13: 34839921 - 34841605 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114288.2 NP_001107760.1 3 - 4 264 68
GenBank mRNA AK232217.1 - 3 - 4 777 68
AK345607.1 - 2 - 3 716 68
AK348614.1 - 3 - 4 642 68
AY610403.1 - 1 - 2 497 68
EU352797.1 - 3 - 4 265 68

View Full Product Details