Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCAS4
Entrez Gene ID
100141422
Gene Name
breast carcinoma amplified sequence 4
Gene Aliases
-
Chromosome Location
Chr.17: 52239359 - 52304259 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123082.1 NP_001116554.1 1 - 2 198 77
XM_021077205.1 XP_020932864.1 - 238 77
GenBank mRNA AK392477.1 - 1 - 2 135 77

View Full Product Details