Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTSZ
Entrez Gene ID
100141405
Gene Name
cathepsin Z
Gene Aliases
-
Chromosome Location
Chr.17: 59141497 - 59149377 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123104.1 NP_001116576.1 1 - 2 272 145
GenBank mRNA AK234297.1 - 1 - 2 233 145
AK237527.1 - 1 - 2 198 145

View Full Product Details