Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NCR1
Entrez Gene ID
100141419
Gene Name
natural cytotoxicity triggering receptor 1
Gene Aliases
-
Chromosome Location
Chr.6: 59092258 - 59099992 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123143.1 NP_001116615.1 4 - 5 659 66

View Full Product Details