Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAB22A
Entrez Gene ID
100144535
Gene Name
RAB22A, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.17: 58514985 - 58572421 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123174.1 NP_001116646.1 5 - 6 626 70

View Full Product Details