Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TFAP2C
Entrez Gene ID
100144525
Gene Name
transcription factor AP-2 gamma
Gene Aliases
-
Chromosome Location
Chr.17: 57164400 - 57174923 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123201.1 NP_001116673.1 6 - 7 1308 85

View Full Product Details