Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD1
Entrez Gene ID
396959
Gene Name
ankyrin repeat domain 1
Gene Aliases
CARP
Chromosome Location
Chr.14: 102603777 - 102612857 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213922.1 NP_999087.1 4 - 5 515 119
XM_021072076.1 XP_020927735.1 - 534 119
GenBank mRNA AY248702.1 - 4 - 5 515 119
AY609421.1 - 4 - 5 508 119
FJ475066.1 - 4 - 5 515 119
FJ475067.1 - 4 - 5 515 119
FJ475068.1 - 4 - 5 515 119

View Full Product Details