Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSMB8
Entrez Gene ID
396975
Gene Name
proteasome 20S subunit beta 8
Gene Aliases
LMP7
Chromosome Location
Chr.7: 25058456 - 25061666 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213935.2 NP_999100.2 4 - 5 668 76
GenBank mRNA AF059493.1 - 4 - 5 493 76
AK237007.1 - 4 - 5 588 76
DB815752.1 - 2 - 3 560 76
DQ917650.1 - 4 - 5 535 76

View Full Product Details