Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
WFDC2
Entrez Gene ID
396987
Gene Name
WAP four-disulfide core domain 2
Gene Aliases
E4
Chromosome Location
Chr.17: 47816452 - 47824087 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213947.1 NP_999112.1 1 - 2 105 77
GenBank mRNA AJ506744.1 - 1 - 2 105 77

View Full Product Details