Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSMC5
Entrez Gene ID
397043
Gene Name
proteasome 26S subunit, ATPase 5
Gene Aliases
TBP10
Chromosome Location
Chr.12: 15101946 - 15106326 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213983.1 NP_999148.1 11 - 12 1313 64
XM_005668681.3 XP_005668738.1 - 1396 64
XM_013980839.2 XP_013836293.1 - 1445 64
XM_021066208.1 XP_020921867.1 - 1489 64
XM_021066209.1 XP_020921868.1 - 1540 64
XM_021066210.1 XP_020921869.1 - 1353 64
XM_021066211.1 XP_020921870.1 - 1364 64
GenBank mRNA AK234194.1 - 11 - 12 1203 64
AK395258.1 - 11 - 12 1225 64
AK396074.1 - 11 - 12 1198 64
X89719.1 - - 1141 64

View Full Product Details