Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSMC5
Entrez Gene ID
397043
Gene Name
proteasome 26S subunit, ATPase 5
Gene Aliases
TBP10
Chromosome Location
Chr.12: 15101946 - 15106326 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213983.1 NP_999148.1 9 - 10 1117 95
XM_005668681.3 XP_005668738.1 - 1200 95
XM_013980839.2 XP_013836293.1 - 1249 95
XM_021066208.1 XP_020921867.1 - 1202 95
XM_021066209.1 XP_020921868.1 - 1253 95
XM_021066210.1 XP_020921869.1 - 1066 95
XM_021066211.1 XP_020921870.1 - 1077 95
GenBank mRNA AK234194.1 - 9 - 10 1007 95
AK395258.1 - 9 - 10 1029 95
AK396074.1 - 9 - 10 1002 95
X89719.1 - - 945 95

View Full Product Details