Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYL1
Entrez Gene ID
397666
Gene Name
myosin light chain 1
Gene Aliases
MLC
Chromosome Location
Chr.15: 113038083 - 113051315 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214374.2 NP_999539.2 5 - 6 507 131
GenBank mRNA AK347117.1 - 5 - 6 667 131
AK347215.1 - 5 - 6 833 131
AK347231.1 - 5 - 6 517 131
AK394612.1 - 6 - 7 522 131
AK394636.1 - 5 - 6 463 131
AK394638.1 - 6 - 7 491 131
AK394642.1 - 5 - 6 676 131
AK394655.1 - 5 - 6 494 131
AK394666.1 - 5 - 6 494 131
AK394667.1 - 5 - 6 497 131
AK394722.1 - 5 - 6 498 131
AK399622.1 - 5 - 6 496 131
AK399655.1 - 5 - 6 494 131
AY680665.1 - 5 - 6 515 131
AY680667.1 - 5 - 6 515 131
AY680668.1 - 5 - 6 515 131
AY680669.1 - 5 - 6 515 131
AY864611.1 - 5 - 6 507 131
AY864615.1 - 6 - 7 537 131
AY864616.1 - 5 - 6 476 131
AY894693.1 - 5 - 6 507 131
DQ629158.1 - 5 - 6 674 131
DQ629159.1 - 5 - 6 496 131
DQ629160.1 - 6 - 7 523 131
GU195782.1 - 6 - 7 524 131
GU195783.1 - 5 - 6 465 131
GU195784.1 - 5 - 6 460 131
X94689.2 - - 477 131
Y18404.1 - - 71 131

View Full Product Details